Assay Details
| Gene Symbol: Naa25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172722.3 | NP_766310.2 | 18 - 19 | 2268 | 91 | |
| GenBank mRNA | AK041906.1 | - | 4 - 5 | 585 | 91 | |
| AK049325.1 | - | 18 - 19 | 2269 | 91 | ||
| AK149758.1 | - | 18 - 19 | 2269 | 91 | ||
| AK165175.1 | - | 18 - 19 | 2260 | 92 | ||
| AK167405.1 | - | 18 - 19 | 2286 | 91 | ||
| AK167491.1 | - | 18 - 19 | 2286 | 91 | ||
| BC028868.1 | - | 16 - 17 | 2048 | 91 | ||
| BC037595.1 | - | 18 - 19 | 2249 | 91 | ||
| BC055480.1 | - | 17 - 18 | 2170 | 91 | ||
| BC082314.1 | - | 18 - 19 | 2254 | 91 | ||
| BC106182.1 | - | 18 - 19 | 2243 | 91 | ||
| BC132172.1 | - | 18 - 19 | 2266 | 40130449 | 91 | |
| BC138093.1 | - | 18 - 19 | 2266 | 8861112 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa25
Genomic Map