Assay Details
| Gene Symbol: Naa25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172722.3 | NP_766310.2 | 2 - 3 | 168 | 134 | |
| GenBank mRNA | AK049325.1 | - | 2 - 3 | 169 | 134 | |
| AK052060.1 | - | 2 - 3 | 156 | 134 | ||
| AK149758.1 | - | 2 - 3 | 169 | 134 | ||
| AK165175.1 | - | 2 - 3 | 159 | 134 | ||
| AK167405.1 | - | 2 - 3 | 186 | 134 | ||
| AK167491.1 | - | 2 - 3 | 186 | 134 | ||
| BC037595.1 | - | 2 - 3 | 149 | 134 | ||
| BC082314.1 | - | 2 - 3 | 154 | 134 | ||
| BC106182.1 | - | 2 - 3 | 143 | 134 | ||
| BC132172.1 | - | 2 - 3 | 166 | 40130449 | 134 | |
| BC138093.1 | - | 2 - 3 | 166 | 8861112 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa25
Genomic Map