Assay Details
| Gene Symbol: Naa25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172722.3 | NP_766310.2 | 7 - 8 | 680 | 66 | |
| GenBank mRNA | AK049325.1 | - | 7 - 8 | 681 | 66 | |
| AK052060.1 | - | 7 - 8 | 668 | 66 | ||
| AK149758.1 | - | 7 - 8 | 681 | 66 | ||
| AK165175.1 | - | 7 - 8 | 671 | 66 | ||
| AK167405.1 | - | 7 - 8 | 698 | 66 | ||
| AK167491.1 | - | 7 - 8 | 698 | 66 | ||
| BC028868.1 | - | 5 - 6 | 460 | 66 | ||
| BC037595.1 | - | 7 - 8 | 661 | 66 | ||
| BC055480.1 | - | 6 - 7 | 582 | 66 | ||
| BC082314.1 | - | 7 - 8 | 666 | 66 | ||
| BC106182.1 | - | 7 - 8 | 655 | 66 | ||
| BC132172.1 | - | 7 - 8 | 678 | 40130449 | 66 | |
| BC138093.1 | - | 7 - 8 | 678 | 8861112 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Naa25
Genomic Map