Assay Details
| Gene Symbol: Glt1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177005.4 | NP_795979.2 | 2 - 3 | 240 | 59 | |
| GenBank mRNA | AK050379.1 | - | 2 - 3 | 241 | 59 | |
| AK077577.1 | - | 2 - 3 | 241 | 59 | ||
| BY337475.1 | - | 2 - 3 | 241 | 59 | ||
More Information
Set Membership: |
|

Glt1d1
Genomic Map