Assay Details
| Gene Symbol: Usp42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029749.2 | NP_084025.2 | 3 - 4 | 572 | 78 | |
| GenBank mRNA | AK010801.1 | - | 2 - 3 | 382 | 78 | |
| AK206149.1 | - | 1 - 2 | 77 | 78 | ||
| AY336567.1 | - | 2 - 3 | 445 | 78 | ||
| BC137852.1 | - | 2 - 3 | 465 | 8860871 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp42
Genomic Map