Assay Details
| Gene Symbol: Usp42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029749.2 | NP_084025.2 | 4 - 5 | 682 | 82 | |
| GenBank mRNA | AK010801.1 | - | 3 - 4 | 492 | 82 | |
| AY336567.1 | - | 3 - 4 | 555 | 82 | ||
| BC137852.1 | - | 3 - 4 | 575 | 8860871 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp42
Genomic Map