Assay Details
| Gene Symbol: Usp42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029749.2 | NP_084025.2 | 9 - 10 | 1112 | 72 | |
| GenBank mRNA | AK010801.1 | - | 8 - 9 | 922 | 72 | |
| AY336567.1 | - | 8 - 9 | 985 | 72 | ||
| BC137852.1 | - | 8 - 9 | 1005 | 8860871 | 72 | |
| BG079818.2 | - | 5 - 6 | 394 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp42
Genomic Map