Assay Details
| Gene Symbol: A630023P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102290.1 | 1 - 2 | 149 | 71 | ||
| GenBank mRNA | AK088579.1 | - | 1 - 2 | 150 | 71 | |
More Information
Updated Mapping Information
Set Membership: | 
		   
					
						
							 | 
				

A630023P12Rik
Genomic Map