Assay Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033202.3 | NP_001028374.1 | 2 - 3 | 290 | 72 | |
| GenBank mRNA | AK144578.1 | - | 2 - 3 | 291 | 72 | |
| BC038606.1 | - | 2 - 3 | 312 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp30
Genomic Map