Assay Details
| Gene Symbol: Usp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033202.3 | NP_001028374.1 | 5 - 6 | 680 | 62 | |
| GenBank mRNA | AK078164.1 | - | 1 - 2 | 32 | 62 | |
| BC038606.1 | - | 5 - 6 | 702 | 62 | ||
More Information
Set Membership: |
|

Usp30
Genomic Map