Assay Details
| Gene Symbol: Zfp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145916.2 | NP_666021.2 | 2 - 3 | 121 | 102 | |
| GenBank mRNA | AK168948.1 | - | 2 - 3 | 76 | 102 | |
| BC011501.1 | - | 2 - 3 | 139 | 3584546 | 102 | |
| BU613201.1 | - | 2 - 3 | 121 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK168948.1, BC011501.1, NM_145916.2 transcripts.
Set Membership: |
|

Zfp7
Genomic Map