Assay Details
| Gene Symbol: Zfp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144887.1 | - | 2 - 3 | 333 | 106 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144887.1 transcripts.
Set Membership: |
|

Zfp7
Genomic Map