Assay Details
| Gene Symbol: Vegfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009506.2 | NP_033532.1 | 5 - 6 | 1046 | 138 | |
| GenBank mRNA | AK047844.1 | - | 973 | 138 | ||
| BC096377.1 | - | 5 - 6 | 951 | 5002720 | 138 | |
| U58112.1 | - | 5 - 6 | 942 | 138 | ||
| U73620.1 | - | 5 - 6 | 952 | 138 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vegfc
Genomic Map