Assay Details
| Gene Symbol: Slc6a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146013.1 | NP_001139485.1 | 10 - 11 | 1715 | 70 | |
| NM_148931.3 | NP_683733.2 | 9 - 10 | 1633 | 70 | ||
| GenBank mRNA | AF411042.3 | - | 10 - 11 | 1769 | 70 | |
| AY147186.2 | - | 10 - 11 | 1868 | 70 | ||
| BC138125.1 | - | 9 - 10 | 1636 | 8861144 | 70 | |
| BC138126.1 | - | 9 - 10 | 1636 | 8861145 | 70 | |
| BC157997.1 | - | 10 - 11 | 1715 | 9007308 | 70 | |
| BC172125.1 | - | 9 - 10 | 1576 | 70 | ||
More Information
Set Membership: |
|

Slc6a5
Genomic Map