Assay Details
| Gene Symbol: Slc6a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146013.1 | NP_001139485.1 | 2 - 3 | 626 | 114 | |
| NM_148931.3 | NP_683733.2 | 1 - 2 | 544 | 114 | ||
| GenBank mRNA | AB118159.1 | - | 2 - 3 | 573 | 114 | |
| AF411042.3 | - | 2 - 3 | 680 | 114 | ||
| AY147186.2 | - | 2 - 3 | 779 | 114 | ||
| BC138125.1 | - | 1 - 2 | 547 | 8861144 | 114 | |
| BC138126.1 | - | 1 - 2 | 547 | 8861145 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a5
Genomic Map