Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700008183.mm.1 |
TC0700000907.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700008183.mm.1 |
NM_001146013.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008183.mm.1 |
NM_001146013.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008183.mm.1 |
NM_148931.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008183.mm.1 |
NM_148931.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008182.mm.1 |
TC0700000907.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700008182.mm.1 |
NM_001146013.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008182.mm.1 |
NM_001146013.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008182.mm.1 |
NM_148931.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700008182.mm.1 |
NM_148931.3 |
Refseq |