Assay Details
| Gene Symbol: Lipk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205349.1 | NP_001192278.1 | 1 - 2 | 130 | 66 | |
| NM_172837.4 | NP_766425.1 | 1 - 2 | 130 | 66 | ||
| GenBank mRNA | AK037106.1 | - | 1 - 2 | 95 | 66 | |
| AK162706.1 | - | 1 - 2 | 52 | 66 | ||
| BY150606.1 | - | 1 - 2 | 130 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK037106.1, AK162706.1, NM_001205349.1, NM_172837.4 transcripts.
Set Membership: |
|

Lipk
Genomic Map