Assay Details
| Gene Symbol: Saa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011316.3 | NP_035446.1 | 2 - 3 | 205 | 70 | |
| GenBank mRNA | BC019212.1 | - | 2 - 3 | 241 | 5053373 | 70 |
| U02554.1 | - | 2 - 3 | 236 | 70 | ||
More Information
Set Membership: |
|

Saa4
Genomic Map