Assay Details
| Gene Symbol: Saa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011316.3 | NP_035446.1 | 3 - 4 | 350 | 61 | |
| GenBank mRNA | BC019212.1 | - | 3 - 4 | 386 | 5053373 | 61 |
| U02554.1 | - | 3 - 4 | 381 | 61 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P404329 | NM_011316.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P404329 | BC019212.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P404329 | NM_011316.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P404329 | BC019212.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684861 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168358 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88683 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581573 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 94333 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116072 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P404329 | NM_011316.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P404329 | BC019212.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P404329 | NM_011316.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P404329 | BC019212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P142529 | NM_011316.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P142529 | BC019212.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P142529 | U02554.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100937 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100947 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100965 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100953 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100957 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100961 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100955 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25100991 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06186631 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116072 | - | - |
| Whole Mouse Genome G4122A | A_52_P404329 | NM_011316.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P404329 | BC019212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P404329 | NM_011316.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P404329 | BC019212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P142529 | NM_011316.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P142529 | BC019212.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P142529 | U02554.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135692 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116072 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06557703 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06557701 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06365098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100894 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100890 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25100928 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06335192 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05924900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06586403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05924912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100955 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06287550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06335195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05924907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06287551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06557703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06156441 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100948 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100951 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06184057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06416478 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05924908 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100944 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06450766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06414098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06365098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06459191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100946 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05931117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100942 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06300624 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100972 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25100987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06414100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06335193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06641257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05931122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05931112 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10563605 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563604 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563603 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563606 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10563607 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419318_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419319_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10563602 | - | - |
| Mouse Genome 430 2.0 Array | 1419318_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419318_at | - | - |
| Mouse Genome 430 2.0 Array | 1419319_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419319_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20211_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20211_at | - | - |
| Mouse Expression Set 430 | 1419318_PM_at | - | - |
| Mouse Expression Set 430 | 1419319_at | - | - |
| Mouse Expression Set 430 | 1419318_at | - | - |
| Mouse Expression Set 430 | 1419319_PM_at | - | - |
| Murine Genome U74v2 Set | 92242_at | - | - |
| Murine Genome U74v2 Set | 92242_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419318_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419319_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419318_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419319_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419319_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419318_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419319_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419318_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 598592 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 430706 | AK037670.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 430706 | AK151968.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 430706 | AK152103.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 430706 | AK160006.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 430706 | AK166466.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 430706 | AK170264.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 430706 | BC019207.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 430706 | BC053041.1 | genbank_mrna_mouse |
| MouseWG-6 v2 Expression BeadChip | 430706 | NM_010700.2 | refseq_mrna_mouse_NM |
| MouseWG-6 v2 Expression BeadChip | 430706 | X64414.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | AK037670.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | AK151968.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | AK152103.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | AK160006.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | AK166466.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | AK170264.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | BC019207.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | BC053041.1 | genbank_mrna_mouse |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | NM_010700.2 | refseq_mrna_mouse_NM |
| Illumina mouse-6 v1.0 expression beadchip | 430706 | X64414.1 | genbank_mrna_mouse |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000430706 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1247506 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1247506 | - | - |
| mouse-6 v1.1 (Illumina) | scl31342.5.1_38-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | AK037670.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | AK151968.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | AK152103.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | AK160006.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | AK166466.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | AK170264.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | BC019207.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | BC053041.1 | genbank_mrna_mouse |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | NM_010700.2 | refseq_mrna_mouse_NM |
| Illumina Sentrix mouse-6.v1 bead array | 430706 | X64414.1 | genbank_mrna_mouse |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31342.5.1_38-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | AK037670.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | AK151968.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | AK152103.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | AK160006.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | AK166466.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | AK170264.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | BC019207.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | BC053041.1 | genbank_mrna_mouse |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | NM_010700.2 | refseq_mrna_mouse_NM |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 430706 | X64414.1 | genbank_mrna_mouse |
| MouseRef-8 v2 Expression BeadChip | ILMN_1247506 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1247506 | - | - |

Saa4
Genomic Map