Assay Details
| Gene Symbol: Abcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018795.2 | NP_061265.2 | 11 - 12 | 1431 | 83 | |
| GenBank mRNA | AB028737.1 | - | 11 - 12 | 1437 | 83 | |
| BC040400.1 | - | 11 - 12 | 1439 | 83 | ||
| EF109740.1 | - | 11 - 12 | 1431 | 83 | ||
| EF109741.1 | - | 11 - 12 | 1431 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Abcc6
Genomic Map