Assay Details
| Gene Symbol: Abcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018795.2 | NP_061265.2 | 1 - 2 | 60 | 104 | |
| GenBank mRNA | BC040400.1 | - | 1 - 2 | 68 | 104 | |
| EF109740.1 | - | 1 - 2 | 60 | 104 | ||
| EF109741.1 | - | 1 - 2 | 60 | 104 | ||
More Information
Set Membership: |
|

Abcc6
Genomic Map