Assay Details
| Gene Symbol: Abcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB028737.1 | - | 21 - 22 | 2783 | 70 | |
| EF109740.1 | - | 21 - 22 | 2777 | 70 | ||
| EF109741.1 | - | 21 - 22 | 2777 | 70 | ||
More Information
Set Membership: |
|

Abcc6
Genomic Map