Assay Details
| Gene Symbol: Rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133914.3 | NP_598675.2 | 13 - 14 | 1570 | 80 | |
| GenBank mRNA | AK044762.1 | - | 11 - 12 | 1515 | 80 | |
| AK133565.1 | - | 12 - 13 | 1719 | 80 | ||
| AK154335.1 | - | 13 - 14 | 1571 | 80 | ||
| AK155146.1 | - | 13 - 14 | 1571 | 80 | ||
| AK220218.1 | - | 13 - 14 | 1446 | 80 | ||
| AY591339.1 | - | 13 - 14 | 1471 | 80 | ||
| BC027230.1 | - | 12 - 13 | 1691 | 80 | ||
| BC049381.1 | - | 12 - 13 | 1775 | 80 | ||
| BC057460.1 | - | 13 - 14 | 1424 | 4527934 | 80 | |
More Information
Set Membership: |
|

Rasa4
Genomic Map