Assay Details
| Gene Symbol: Rasa4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039103.3 | NP_001034192.1 | 4 - 5 | 493 | 92 | |
| NM_133914.3 | NP_598675.2 | 4 - 5 | 493 | 92 | ||
| GenBank mRNA | AK014220.1 | - | 4 - 5 | 412 | 92 | |
| AK044762.1 | - | 4 - 5 | 321 | 92 | ||
| AK133565.1 | - | 4 - 5 | 412 | 92 | ||
| AK154335.1 | - | 4 - 5 | 494 | 92 | ||
| AK155146.1 | - | 4 - 5 | 494 | 92 | ||
| AK220218.1 | - | 4 - 5 | 369 | 92 | ||
| AY591339.1 | - | 4 - 5 | 394 | 92 | ||
| BC049381.1 | - | 4 - 5 | 468 | 92 | ||
| BC057460.1 | - | 4 - 5 | 347 | 4527934 | 92 | |
| DQ317932.1 | - | 4 - 5 | 377 | 92 | ||
More Information
Set Membership: |
|

Rasa4
Genomic Map