Assay Details
| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172405.3 | NP_765993.1 | 5 - 6 | 486 | 83 | |
| GenBank mRNA | AK014420.1 | - | 5 - 6 | 512 | 83 | |
| AK042001.1 | - | 5 - 6 | 495 | 83 | ||
| AK042339.1 | - | 5 - 6 | 495 | 83 | ||
| AK051816.1 | - | 5 - 6 | 487 | 83 | ||
| AK167540.1 | - | 5 - 6 | 517 | 83 | ||
| BC029845.1 | - | 5 - 6 | 491 | 4010744 | 83 | |
| BC043923.1 | - | 4 - 5 | 400 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fam175a
Genomic Map