Assay Details
| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014420.1 | - | 7 - 8 | 725 | 131 | |
| AK042001.1 | - | 7 - 8 | 708 | 131 | ||
| AK042339.1 | - | 7 - 8 | 708 | 131 | ||
More Information
Set Membership: |
|

Fam175a
Genomic Map