Assay Details
| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042001.1 | - | 8 - 9 | 770 | 140 | |
| AK042339.1 | - | 8 - 9 | 770 | 140 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam175a| Gene Symbol: Fam175a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042001.1 | - | 8 - 9 | 770 | 140 | |
| AK042339.1 | - | 8 - 9 | 770 | 140 | ||
Set Membership: |
|