Assay Details
Gene Symbol: Rasgef1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145839.2 | NP_665838.1 | 5 - 6 | 876 | 82 | |
NM_181318.4 | NP_851835.1 | 5 - 6 | 873 | 82 | ||
GenBank mRNA | AK028756.1 | - | 5 - 6 | 765 | 82 | |
AK090085.1 | - | 5 - 6 | 768 | 82 | ||
AK149812.1 | - | 5 - 6 | 795 | 82 | ||
AY129963.1 | - | 5 - 6 | 803 | 82 | ||
BC050858.1 | - | 5 - 6 | 806 | 6517502 | 82 | |
BC052726.1 | - | 5 - 6 | 767 | 6813357 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
