Assay Details
| Gene Symbol: Ptp4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166389.1 | NP_001159861.1 | 1 - 2 | 369 | 63 | |
| GenBank mRNA | CA538895.1 | - | 1 - 2 | 286 | 63 | |
| CK620602.1 | - | 1 - 2 | 369 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001166389.1 transcripts.
Set Membership: |
|

Ptp4a3
Genomic Map