Assay Details
| Gene Symbol: Ptp4a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166388.1 | NP_001159860.1 | 1 - 2 | 641 | 66 | |
| GenBank mRNA | BC066043.1 | - | 1 - 2 | 641 | 6415021 | 66 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC066043.1, NM_001166388.1 transcripts.
Set Membership: |
|

Ptp4a3
Genomic Map