Assay Details
| Gene Symbol: Sla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009192.3 | NP_033218.1 | 2 - 3 | 526 | 88 | |
| GenBank mRNA | AJ131777.1 | - | 1 - 2 | 164 | 88 | |
| AK037901.1 | - | 1 - 2 | 171 | 88 | ||
| AK137954.1 | - | 1 - 2 | 192 | 88 | ||
| BC032922.1 | - | 1 - 2 | 62 | 1430090 | 88 | |
| CD578683.1 | - | 2 - 3 | 343 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ131777.1, AK037901.1, AK137954.1, BC032922.1, NM_009192.3 transcripts.
Set Membership: |
|

Sla
Genomic Map