Assay Details
| Gene Symbol: Cdc37l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK012312.1 | - | 5 - 6 | 787 | 130 | |
| AK016651.1 | - | 5 - 6 | 930 | 130 | ||
| AK038578.1 | - | 5 - 6 | 752 | 130 | ||
| AK044291.1 | - | 5 - 6 | 900 | 130 | ||
| AK048044.1 | - | 5 - 6 | 900 | 130 | ||
| AK076231.1 | - | 5 - 6 | 914 | 130 | ||
| AK078139.1 | - | 5 - 6 | 898 | 130 | ||
| AK078468.1 | - | 5 - 6 | 914 | 130 | ||
| AK078872.1 | - | 5 - 6 | 834 | 130 | ||
| AK079556.1 | - | 5 - 6 | 751 | 130 | ||
| AK161465.1 | - | 5 - 6 | 809 | 130 | ||
| AK165398.1 | - | 5 - 6 | 916 | 130 | ||
| BC031761.1 | - | 5 - 6 | 780 | 4235549 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdc37l1
Genomic Map