Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700006069.mm.1 |
TC0700000756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006069.mm.1 |
NM_001271019.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006069.mm.1 |
NM_145581.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006078.mm.1 |
TC0700000756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006078.mm.1 |
NM_001271019.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006078.mm.1 |
NM_145581.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006080.mm.1 |
TC0700000756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006080.mm.1 |
NM_001271019.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006080.mm.1 |
NM_145581.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006081.mm.1 |
TC0700000756.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006081.mm.1 |
NM_001271019.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006081.mm.1 |
NM_145581.2 |
Refseq |