Assay Details
Gene Symbol: Siglecf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271019.1 | NP_001257948.1 | 4 - 5 | 785 | 72 | |
NM_145581.2 | NP_663556.1 | 4 - 5 | 785 | 72 | ||
GenBank mRNA | AF293371.1 | - | 4 - 5 | 721 | 72 | |
AK087658.1 | - | 3 - 4 | 848 | 72 | ||
AK146203.1 | - | 3 - 4 | 822 | 72 | ||
AK155469.1 | - | 4 - 5 | 732 | 72 | ||
AY210401.1 | - | 3 - 4 | 681 | 72 | ||
BC145038.1 | - | 3 - 4 | 716 | 9053586 | 72 | |
BC145987.1 | - | 3 - 4 | 716 | 40131290 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
