Assay Details
Gene Symbol: Siglecf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145581.2 | NP_663556.1 | 7 - 8 | 1385 | 60 | |
GenBank mRNA | AF293371.1 | - | 7 - 8 | 1321 | 60 | |
AK087658.1 | - | 6 - 7 | 1448 | 60 | ||
AK146203.1 | - | 6 - 7 | 1422 | 60 | ||
AK155469.1 | - | 7 - 8 | 1334 | 60 | ||
AY210401.1 | - | 6 - 7 | 1281 | 60 | ||
BC145987.1 | - | 6 - 7 | 1316 | 40131290 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
