Assay Details
| Gene Symbol: Ptov1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133949.1 | NP_598710.1 | 10 - 11 | 1306 | 66 | |
| GenBank mRNA | AK003974.1 | - | 1 - 2 | 67 | 66 | |
| BC008543.1 | - | 10 - 11 | 1182 | 3496037 | 66 | |
| BC024632.1 | - | 10 - 11 | 1306 | 4222241 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptov1
Genomic Map