Assay Details
| Gene Symbol: Snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005045.2 | - | 7 - 8 | 670 | 81 | |
| AK012272.1 | - | 5 - 6 | 319 | 81 | ||
| AK077425.1 | - | 7 - 8 | 672 | 81 | ||
| BC049128.1 | - | 7 - 8 | 636 | 81 | ||
More Information
Set Membership: |
|

Snrnp70
Genomic Map