Assay Details
| Gene Symbol: Gm16226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK160425.1 | - | 115 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 38691 |
Gm38691 | Sense | Chr. 5 - 99681301 - 99690324 on NCBI Build GRCm38 | ||
| Downstream |
predicted gene, 38413 |
Gm38413 | A830083F22 | 27.3kb | Anti-sense | Chr. 5 - 99730841 - 99754303 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm16226
Genomic Map