Assay Details
| Gene Symbol: Mrps18c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026826.1 | NP_081102.1 | 2 - 3 | 157 | 110 | |
| GenBank mRNA | AK088290.1 | - | 2 - 3 | 170 | 110 | |
| AK145935.1 | - | 2 - 3 | 174 | 110 | ||
| BC028442.1 | - | 2 - 3 | 157 | 1196057 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mrps18c
Genomic Map