Assay Details
| Gene Symbol: Antxr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133738.1 | NP_598499.1 | 12 - 13 | 1450 | 82 | |
| GenBank mRNA | AK155460.1 | - | 12 - 13 | 1533 | 82 | |
| AK166124.1 | - | 1222 | 82 | |||
| AK170655.1 | - | 12 - 13 | 1088 | 82 | ||
| AK171292.1 | - | 12 - 13 | 1125 | 82 | ||
| BC003908.1 | - | 3 - 4 | 240 | 82 | ||
| BC076595.1 | - | 12 - 13 | 1450 | 30649897 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Antxr2
Genomic Map