Assay Details
| Gene Symbol: Paqr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198422.2 | NP_940814.1 | 2 - 3 | 502 | 76 | |
| GenBank mRNA | AK149213.1 | - | 2 - 3 | 481 | 76 | |
| AK172689.1 | - | 2 - 3 | 457 | 76 | ||
| AY424292.1 | - | 2 - 3 | 502 | 76 | ||
| BC050248.1 | - | 2 - 3 | 502 | 76 | ||
| BC051173.1 | - | 2 - 3 | 502 | 76 | ||
| BC120853.1 | - | 2 - 3 | 487 | 40129776 | 76 | |
| BC120855.1 | - | 2 - 3 | 488 | 40129778 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Paqr3
Genomic Map