Assay Details
| Gene Symbol: Adgrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081342.1 | NP_001074811.1 | 16 - 17 | 2173 | 82 | |
| GenBank mRNA | AK153762.1 | - | 16 - 17 | 2175 | 82 | |
| AY255618.1 | - | 1 - 2 | 58 | 82 | ||
| BC151161.1 | - | 16 - 17 | 1875 | 9088050 | 82 | |
| BC157994.1 | - | 15 - 16 | 1779 | 9007307 | 82 | |
| BC158001.1 | - | 15 - 16 | 1779 | 9007310 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adgrd1
Genomic Map