Assay Details
Gene Symbol: Zfp707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005330.1 | - | 1 - 2 | 132 | 57 | |
AK137962.1 | - | 1 - 2 | 253 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
