| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1500003912.mm.1 |
NM_001081065.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500003912.mm.1 |
NM_001081065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500003912.mm.1 |
TC1500000568.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500003913.mm.1 |
NM_001081065.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500003913.mm.1 |
NM_001081065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500003913.mm.1 |
TC1500000568.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500003918.mm.1 |
NM_001081065.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500003918.mm.1 |
NM_001081065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500003918.mm.1 |
TC1500000568.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500003911.mm.1 |
NM_001081065.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500003911.mm.1 |
NM_001081065.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500003911.mm.1 |
TC1500000568.mm.1 |
MTA TC |