Assay Details
| Gene Symbol: Pprc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081214.1 | NP_001074683.1 | 2 - 3 | 376 | 80 | |
| GenBank mRNA | AK044522.1 | - | 2 - 3 | 335 | 80 | |
| AK191373.1 | - | 1 - 2 | 179 | 80 | ||
| BC066048.1 | - | 2 - 3 | 330 | 6825158 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pprc1
Genomic Map