Assay Details
| Gene Symbol: Ldhc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013580.4 | NP_038608.1 | 2 - 3 | 174 | 64 | |
| GenBank mRNA | AK007058.1 | - | 2 - 3 | 175 | 64 | |
| AK161399.1 | - | 2 - 3 | 162 | 64 | ||
| BC049602.1 | - | 2 - 3 | 178 | 6774010 | 64 | |
| JQ173119.1 | - | 2 - 3 | 151 | 64 | ||
| JQ173120.1 | - | 2 - 3 | 151 | 64 | ||
| X04752.1 | - | 2 - 3 | 182 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldhc
Genomic Map