Assay Details
| Gene Symbol: Acads | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007383.3 | NP_031409.2 | 5 - 6 | 697 | 121 | |
| GenBank mRNA | AK155361.1 | - | 5 - 6 | 676 | 121 | |
| AK169428.1 | - | 5 - 6 | 634 | 121 | ||
| BC016259.1 | - | 5 - 6 | 664 | 4913813 | 121 | |
| L11163.1 | - | 5 - 6 | 621 | 121 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acads
Genomic Map