Assay Details
| Gene Symbol: Acads | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007383.3 | NP_031409.2 | 8 - 9 | 1106 | 72 | |
| GenBank mRNA | AK155361.1 | - | 8 - 9 | 1085 | 72 | |
| AK169428.1 | - | 8 - 9 | 1043 | 72 | ||
| BC016259.1 | - | 8 - 9 | 1073 | 4913813 | 72 | |
| BC055063.1 | - | 7 - 8 | 1259 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acads
Genomic Map