Assay Details
| Gene Symbol: Msi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008629.1 | NP_032655.1 | 5 - 6 | 377 | 88 | |
| GenBank mRNA | AK084019.1 | - | 5 - 6 | 360 | 88 | |
| D49654.1 | - | 5 - 6 | 377 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Msi1
Genomic Map