Assay Details
| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133906.4 | NP_598667.2 | 3 - 4 | 809 | 82 | |
| GenBank mRNA | AK020039.1 | - | 3 - 4 | 785 | 82 | |
| AK028932.1 | - | 3 - 4 | 740 | 82 | ||
| AK031759.1 | - | 3 - 4 | 790 | 82 | ||
| AK036807.1 | - | 3 - 4 | 792 | 82 | ||
| AK054323.1 | - | 3 - 4 | 780 | 82 | ||
| AK135999.1 | - | 3 - 4 | 759 | 82 | ||
| AK154831.1 | - | 3 - 4 | 789 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zkscan1
Genomic Map